3-Fluor-4-iodtoluol

Lieferant: CHEMPUR

FL13321.POR 013321-1G 013321-5G
CHMPFL13321.POREA 0 EUR
CHMPFL13321.POR FLUO013321-1G FLUO013321-5G
3-Fluor-4-iodtoluol
3-Fluor-4-iodtoluol
Formel: C₇H₆FI
MDL: MFCD00143162
CAS-Nummer: 452-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR