3-Fluor-4-iodtoluol

Lieferant: FLUOROCHEM

Synonyme: 1-Fluor-2-iod-5-methylbenzol

021397-5G 021397-1G
FLUO021397-5GEA 0 EUR
FLUO021397-5G FLUO021397-1G
3-Fluor-4-iodtoluol
3-Fluor-4-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD00143162
CAS-Nummer: 452-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR