3-Fluor-4-iodtoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Fluor-2-iod-5-methylbenzol

M-Clarity™ Qualitätsstufe = MQ100

635154-1G 635154-5G
SIAL635154-1GEA 39.5 EUR
SIAL635154-1G SIAL635154-5G
3-Fluor-4-iodtoluol, Sigma-Aldrich®
3-Fluor-4-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD00143162
CAS-Nummer: 452-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR