2-Fluor-6-iodtoluol

Lieferant: CHEMPUR

FL13320.POR 013320-25G 013320-5G
CHMPFL13320.POREA 0 EUR
CHMPFL13320.POR FLUO013320-25G FLUO013320-5G
2-Fluor-6-iodtoluol
2-Fluor-6-iodtoluol
Formel: C₇H₆FI
MDL: MFCD00142922
CAS-Nummer: 443-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR