5-Fluor-2-iodtoluol

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-6-iodtoluol, 1-Fluor-4-iod-3-methylbenzol

005416-1G 005416-5G 005416-25G
FLUO005416-1GEA 31.1 EUR
FLUO005416-1G FLUO005416-5G FLUO005416-25G
5-Fluor-2-iodtoluol
5-Fluor-2-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236,03 g/mol
MDL: MFCD00039397
CAS-Nummer: 66256-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR