α,α,α-Trifluor-o-tolylisocyanat
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Trifluormethylphenyl)isocyanat
SB01785.1
SB01785.10
MAYBSB01785.1EA
0
EUR
MAYBSB01785.1
MAYBSB01785.10
α,α,α-Trifluor-o-tolylisocyanat
α,α,α-Trifluor-o-tolylisocyanat
Formel:
C₈H₄F₃NO Molecular Weight: 187,12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00002008 CAS-Nummer: 2285-12-3 EINECS: 218-925-5 UN: 2285 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...