2-(Trifluormethyl)phenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: α,α,α-Trifluoro-o-tolyl isothiocyanate

SB01593.1 SB01593EA
MAYBSB01593.1EA 0 EUR
MAYBSB01593.1 MAYBSB01593EA
2-(Trifluormethyl)phenylisothiocyanat
2-(Trifluormethyl)phenylisothiocyanat
Formel: C₈H₄F₃NS
Molecular Weight: 203.19 g/mol
Siedepunkt: 214…216 °C (1013 hPa)
Dichte: 1.346 g/cm³ (20 °C)
MDL: MFCD00039644
CAS-Nummer: 1743-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR