3,4,5-Trifluorbenzylamin
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
(3,4,5-Trifluorophenyl)methanamine
007967-1G
007967-5G
FLUO007967-1GEA
45.9
EUR
FLUO007967-1G
FLUO007967-5G
3,4,5-Trifluorbenzylamin
3,4,5-Trifluorbenzylamin
Formel:
C₇H₆F₃N Molecular Weight: 161.13 g/mol Siedepunkt: 169 Schmelzpunkt: 44 |
MDL:
MFCD00236318 CAS-Nummer: 235088-69-4 UN: 2735 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...