4-Fluorbenzylamin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(4-Fluorophenyl)methanamine
A15486.09
A15486.18
A15486.30
A15486.09EA
29.4
EUR
A15486.09
A15486.18
A15486.30
4-Fluorbenzylamin ≥98%
4-Fluorbenzylamin
Purity: 98+%
CAS No.: 140-75-0
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 66°(150°F)
Refractive Index: 1.5130
Odour: Amine-like
Boiling Point: 182-183°
Beilstein Reference: 636501
UN No.: 2735
MDL No.: MFCD00008120
Formel:
C₇H₈FN Molecular Weight: 125,15 g/mol Siedepunkt: 182…183 °C Dichte: 1,095 Flammpunkt: 66 °C (150 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008120 CAS-Nummer: 140-75-0 EINECS: 205-430-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...