2-Fluor-3-(trifluormethyl)anilin

Lieferant: FLUOROCHEM

Synonyme: 3-Amino-α,α,α,2-tetrafluortoluol, α,α,α,2-Tetrafluor-m-toluidin

CFF629.5 003128-5G 003128-25G
CHMPCFF629.5EA 666 EUR
CHMPCFF629.5 FLUO003128-5G FLUO003128-25G
2-Fluor-3-(trifluormethyl)anilin
2-Fluor-3-(trifluormethyl)anilin
Formel: C₇H₅F₄N
Molecular Weight: 179.12 g/mol
Siedepunkt: 189
Dichte: 1.39 g/cm³ (20 °C)
MDL: MFCD00061241
CAS-Nummer: 123973-25-1
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR