3-Fluor-5-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
Synonyme:
3-Amino-α,α,α,5-tetrafluortoluol, α,α,α,5-Tetrafluor-m-toluidin
003983-1G
003983-5G
003983.25G
FLUO003983-1GEA
31.1
EUR
FLUO003983-1G
FLUO003983-5G
FLUO003983.25G
3-Fluor-5-(trifluormethyl)anilin
3-Fluor-5-(trifluormethyl)anilin
Formel:
C₇H₅F₄N Molecular Weight: 179.12 g/mol |
MDL:
MFCD00061315 CAS-Nummer: 454-67-1 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...