4-Fluor-o-toluidin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Fluor-2-methylanilin, 2-Amino-5-fluortoluol
A11609.06
A11609.14
A11609.06EA
34.2
EUR
A11609.06
A11609.14
4-Fluor-o-toluidin ≥97%
4-Fluor-o-toluidin
Purity: 97%
CAS No.: 452-71-1
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 87°(188°F)
Refractive Index: 1.5370
Odour: Characteristic
Boiling Point: 90-92°/16mm
Beilstein Reference: 1931798
UN No.: 2810
MDL No.: MFCD00007832
Formel:
C₇H₈FN Molecular Weight: 125.15 g/mol Siedepunkt: 90…92 °C (16 mmHg) Schmelzpunkt: 14 Dichte: 1.124 Flammpunkt: 87 °C (188 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007832 CAS-Nummer: 452-71-1 EINECS: 207-208-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...