3-Nitro-5-(trifluormethyl)anilin

Lieferant: CHEMPUR

FL13752.POR 013752-1G 013752-5G
CHMPFL13752.POREA 0 EUR
CHMPFL13752.POR FLUO013752-1G FLUO013752-5G
3-Nitro-5-(trifluormethyl)anilin
3-Nitro-5-(trifluormethyl)anilin
Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206,12 g/mol
MDL: MFCD00061229
CAS-Nummer: 401-94-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR