4-Nitro-3-(trifluormethyl)anilin ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5-Amino-2-nitro-α,α,α-trifluortoluol, 4-Nitro-α,α,α-trifluor-m-toluidin

A13798.03 A13798.06
A13798.03EA 31.9 EUR
A13798.03 A13798.06
4-Nitro-3-(trifluormethyl)anilin ≥98%
4-Nitro-3-(trifluormethyl)anilin

Purity: 98% CAS No.: 393-11-3 Molecular Formula: C7H5F3N2O2 Formula Weight: 206.13 Melting Point: 125-128° Beilstein Reference: 2650702 UN No.: 2811 MDL No.: MFCD00014717

Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 125…128 °C
Dichte: 1,752 g/cm³ (–123.15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014717
CAS-Nummer: 393-11-3
EINECS: 206-884-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR