2,6-Dinitro-4-(trifluormethyl)anilin

Lieferant: BIONET RESEARCH
Achtung

Synonyme: 4-Amino-3,5-dinitro-α,α,α-trifluortoluol, 2,6-Dinitro-α,α,α-trifluor-p-toluidin

1C-124.500 1C-124.1 1C-124.5 1C-124.10
BIOT1C-124.500EA 0 EUR
BIOT1C-124.500 BIOT1C-124.1 BIOT1C-124.5 BIOT1C-124.10
2,6-Dinitro-4-(trifluormethyl)anilin
2,6-Dinitro-4-(trifluormethyl)anilin
Formel: C₇H₄F₃N₃O₄
Molecular Weight: 251.12 g/mol
MDL: MFCD00024260
CAS-Nummer: 445-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR