2-(Trifluormethoxy)benzylamin
Lieferant: FLUOROCHEM
|
Gefahr
|
007550-1G
007550-5G
FLUO007550-1GEA
42.9
EUR
FLUO007550-1G
FLUO007550-5G
2-(Trifluormethoxy)benzylamin
2-(Trifluormethoxy)benzylamin
Formel:
C₈H₈F₃NO Siedepunkt: 77 °C (12 mmHg) |
MDL:
MFCD00085176 CAS-Nummer: 175205-64-8 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...