4-Fluor-3-nitrotoluol

Lieferant: FLUOROCHEM

003233-100G 003233-25G 003233-5G
FLUO003233-100GEA 171 EUR
FLUO003233-100G FLUO003233-25G FLUO003233-5G
4-Fluor-3-nitrotoluol
4-Fluor-3-nitrotoluol
Formel: C₇H₆FNO₂
MDL: MFCD00007060
CAS-Nummer: 446-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR