2,3-Difluor-4-methylanisol

Lieferant: FLUOROCHEM

005744-1G 005744-5G
FLUO005744-1GEA 91.2 EUR
FLUO005744-1G FLUO005744-5G
2,3-Difluor-4-methylanisol
2,3-Difluor-4-methylanisol
Formel: C₈H₈F₂O
Molecular Weight: 158.15 g/mol
MDL: MFCD01631321
CAS-Nummer: 261763-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR