2-Fluor-4-methylanisol

Lieferant: FLUOROCHEM
Achtung

CFF1129. 009215-2G 009215-10G
CHMPCFF1129.EA 0 EUR
CHMPCFF1129. FLUO009215-2G FLUO009215-10G
2-Fluor-4-methylanisol
2-Fluor-4-methylanisol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
MDL: MFCD00060270
CAS-Nummer: 399-55-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR