5-Fluor-2-methylanisol ≥98%

Lieferant: Thermo Scientific

B25629.03 B25629.06
B25629.03EA 20.3 EUR
B25629.03 B25629.06
5-Fluor-2-methylanisol ≥98%
5-Fluor-2-methylanisol

Purity: 98% CAS No.: 95729-22-9 Molecular Formula: C8H9FO Formula Weight: 140.16 Refractive Index: 1.4940

Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673008
CAS-Nummer: 95729-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR