4-(Trifluormethyl)anisol

Lieferant: CHEMPUR

FL13386.POR 013386-25G
CHMPFL13386.POREA 0 EUR
CHMPFL13386.POR FLUO013386-25G
4-(Trifluormethyl)anisol
4-(Trifluormethyl)anisol
Formel: C₈H₇F₃O
Molecular Weight: 176.14 g/mol
MDL: MFCD03094456
CAS-Nummer: 402-52-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR