2-(Difluormethoxy)benzylalkohol

Lieferant: FLUOROCHEM

007855-5G 007855-10G 007855-1G
FLUO007855-5GEA 296 EUR
FLUO007855-5G FLUO007855-10G FLUO007855-1G
2-(Difluormethoxy)benzylalkohol
2-(Difluormethoxy)benzylalkohol
Formel: C₈H₈F₂O₂
Molecular Weight: 174,15 g/mol
MDL: MFCD00236226
CAS-Nummer: 72768-94-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR