3-(Difluormethoxy)benzylalkohol

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-(Difluoromethoxy)phenyl)methanol

H50345.03 H50345.06
H50345.03EA 167 EUR
H50345.03 H50345.06
3-(Difluormethoxy)benzylalkohol
3-(Difluormethoxy)benzylalkohol

CAS No.: 125903-81-3 Molecular Formula: C8H8F2O2 Formula Weight: 174.15 MDL No.: MFCD00236227

Formel: C₈H₈F₂O₂
Molecular Weight: 174,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00236227
CAS-Nummer: 125903-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR