2,3,5,6-Tetrafluorbenzylalkohol ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(2,3,5,6-Tetrafluorphenyl)methanol
B25162.06
B25162.14
B25162.06EA
36
EUR
B25162.06
B25162.14
2,3,5,6-Tetrafluorbenzylalkohol ≥98%
2,3,5,6-Tetrafluorbenzylalkohol
Purity: 98%
CAS No.: 4084-38-2
Molecular Formula: C7H4F4O
Formula Weight: 180.10
Odour: Recognizable
Melting Point: 34-38°
Boiling Point: 220-225°
MDL No.: MFCD00792428
Formel:
C₇H₄F₄O Molecular Weight: 180,1 g/mol Siedepunkt: 220…225 °C Schmelzpunkt: 34…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792428 CAS-Nummer: 4084-38-2 EINECS: 443-840-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...