2,3,5,6-Tetrafluorbenzylalkohol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
(2,3,5,6-Tetrafluorphenyl)methanol
008104-5G
008104-10G
FLUO008104-5GEA
38.9
EUR
FLUO008104-5G
FLUO008104-10G
2,3,5,6-Tetrafluorbenzylalkohol
2,3,5,6-Tetrafluorbenzylalkohol
Formel:
C₇H₄F₄O Molecular Weight: 180.1 g/mol |
MDL:
MFCD00792428 CAS-Nummer: 4084-38-2 EINECS: 443-840-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...