2,3,6-Trifluorbenzylalkohol

Lieferant: FLUOROCHEM

007499-1G 007499-5G 007499-10G
FLUO007499-1GEA 31.1 EUR
FLUO007499-1G FLUO007499-5G FLUO007499-10G
2,3,6-Trifluorbenzylalkohol
2,3,6-Trifluorbenzylalkohol
Formel: C₇H₅F₃O
Molecular Weight: 162.11 g/mol
MDL: MFCD00061205
CAS-Nummer: 114152-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR