3,4,5-Trifluorbenzylalkohol

Lieferant: FLUOROCHEM

007502-1G 007502-5G 007502.10G
FLUO007502-1GEA 116 EUR
FLUO007502-1G FLUO007502-5G FLUO007502.10G
3,4,5-Trifluorbenzylalkohol
3,4,5-Trifluorbenzylalkohol
Formel: C₇H₅F₃O
Molecular Weight: 162,11 g/mol
MDL: MFCD00083527
CAS-Nummer: 220227-37-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR