2-Fluorbenzylalkohol

Lieferant: Thermo Scientific

Synonyme: (2-Fluorphenyl)methanol

SB01147.250 SB01147.1
MAYBSB01147.250EA 0 EUR
MAYBSB01147.250 MAYBSB01147.1
2-Fluorbenzylalkohol
2-Fluorbenzylalkohol
Formel: C₇H₇FO
Molecular Weight: 126,13 g/mol
Siedepunkt: 200 °C (1013 hPa)
Dichte: 1,17 g/cm³ (20 °C)
MDL: MFCD00004601
CAS-Nummer: 446-51-5
EINECS: 207-170-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR