6-Fluor-m-kresol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
1-Fluor-2-hydroxy-4-methylbenzol, 4-Fluor-3-hydroxytoluol, 2-Fluor-5-methylphenol
CFF1130.
007901-5G
007901-25G
CHMPCFF1130.EA
0
EUR
CHMPCFF1130.
FLUO007901-5G
FLUO007901-25G
6-Fluor-m-kresol
6-Fluor-m-kresol
Formel:
C₇H₇FO Molecular Weight: 126.13 g/mol |
MDL:
MFCD00190101 CAS-Nummer: 63762-79-8 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...