3-Fluorbenzylalkohol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3-Fluorophenyl)methanol

A12509.09 A12509.14 A12509.18
A12509.09EA 34.6 EUR
A12509.09 A12509.14 A12509.18
3-Fluorbenzylalkohol ≥98%
3-Fluorbenzylalkohol

Purity: 98% CAS No.: 456-47-3 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 90°(194°F) Refractive Index: 1.5100 Odour: Characteristic Boiling Point: 201° Beilstein Reference: 2242511 MDL No.: MFCD00004631

Formel: C₇H₇FO
Molecular Weight: 126,13 g/mol
Siedepunkt: 201 °C
Dichte: 1,166
Flammpunkt: 90 °C (194 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004631
CAS-Nummer: 456-47-3
EINECS: 207-265-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR