3-Fluorbenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(3-Fluorophenyl)methanol
002170-25G
002170-100G
FLUO002170-25GEA
55.4
EUR
FLUO002170-25G
FLUO002170-100G
3-Fluorbenzylalkohol
3-Fluorbenzylalkohol
Formel:
C₇H₇FO Molecular Weight: 126,13 g/mol Siedepunkt: 104 °C (1013 hPa) Dichte: 1,16 g/cm³ (20 °C) |
MDL:
MFCD00004631 CAS-Nummer: 456-47-3 EINECS: 207-265-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...