5-Fluor-o-kresol

Lieferant: FLUOROCHEM

Synonyme: 5-Fluor-2-methylphenol

013052-5G 013052-25G
FLUO013052-5GEA 35.3 EUR
FLUO013052-5G FLUO013052-25G
5-Fluor-o-kresol
5-Fluor-o-kresol
Formel: C₇H₇FO
Molecular Weight: 126.13 g/mol
MDL: MFCD00673009
CAS-Nummer: 452-85-7
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR