3-Fluor-p-kresol
Lieferant: CHEMPUR
Synonyme:
1-Fluor-3-hydroxy-6-methylbenzol, 2-Fluor-4-hydroxytoluol, 3-Fluor-4-methylphenol
FL13359.POR
013359-1G
013359-5G
CHMPFL13359.POREA
0
EUR
CHMPFL13359.POR
FLUO013359-1G
FLUO013359-5G
3-Fluor-p-kresol
3-Fluor-p-kresol
Formel:
C₇H₇FO Molecular Weight: 126.13 g/mol |
MDL:
MFCD00143164 CAS-Nummer: 452-78-8 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...