2-(Trifluormethyl)thiophenol

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Mercaptobenzotrifluorid, 2-(Trifluormethyl)benzolthiol, 2-(Trifluormethyl)phenylmercaptan

TL00684DA TL00684EA
MAYBTL00684DAEA 0 EUR
MAYBTL00684DA MAYBTL00684EA
2-(Trifluormethyl)thiophenol
2-(Trifluormethyl)thiophenol
Formel: C₇H₅F₃S
Molecular Weight: 178,18 g/mol
MDL: MFCD00068896
CAS-Nummer: 13333-97-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR