2-(Trifluormethyl)thiophenol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Mercaptobenzotrifluorid, 2-(Trifluormethyl)benzolthiol, 2-(Trifluormethyl)phenylmercaptan

007681-5G 007681-1G
FLUO007681-5GEA 137 EUR
FLUO007681-5G FLUO007681-1G
2-(Trifluormethyl)thiophenol
2-(Trifluormethyl)thiophenol
Formel: C₇H₅F₃S
Molecular Weight: 178,18 g/mol
MDL: MFCD00068896
CAS-Nummer: 13333-97-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR