4-(Trifluormethyl)thiophenol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(Trifluormethyl)phenylmercaptan, 4-Mercaptobenzotrifluorid, 4-(Trifluormethyl)benzolthiol

A12725.03 A12725.06
A12725.03EA 61.1 EUR
A12725.03 A12725.06
4-(Trifluormethyl)thiophenol ≥97%
4-(Trifluormethyl)thiophenol

Purity: 97% CAS No.: 825-83-2 Molecular Formula: C7H5F3S Formula Weight: 178.18 Refractive Index: 1.4920 Odour: Stench Boiling Point: 66-68°/15mm Beilstein Reference: 2045665 UN No.: 2810 MDL No.: MFCD00042320

Formel: C₇H₅F₃S
Molecular Weight: 178.18 g/mol
Siedepunkt: 66…68 °C (15 mmHg)
Dichte: 1.300
Lagertemperatur: Raumtemperatur
MDL: MFCD00042320
CAS-Nummer: 825-83-2
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR