2-(Trifluormethyl)benzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 2-(Trifluormethyl)benzol-1-sulfonylchlorid, 2-(Trifluormethyl)benzolsulfonsäurechlorid, 2-(Trifluormethyl)benzolsulfochlorid

RF02653.250 RF02653.1
CHMPRF02653.250EA 0 EUR
CHMPRF02653.250 MAYBRF02653.1
2-(Trifluormethyl)benzolsulfonylchlorid
2-(Trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₄ClF₃O₂S
Molecular Weight: 244,62 g/mol
Siedepunkt: 123…126 °C (1013 hPa)
MDL: MFCD00051696
CAS-Nummer: 776-04-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR