2-(Trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-(Trifluormethyl)benzol-1-sulfonylchlorid, 2-(Trifluormethyl)benzolsulfonsäurechlorid, 2-(Trifluormethyl)benzolsulfochlorid
007587-5G
007587-25G
007587-100G
FLUO007587-5GEA
55.4
EUR
FLUO007587-5G
FLUO007587-25G
FLUO007587-100G
2-(Trifluormethyl)benzolsulfonylchlorid
2-(Trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₄ClF₃O₂S Siedepunkt: 123…126 °C (1013 hPa) |
MDL:
MFCD00051696 CAS-Nummer: 776-04-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...