3-(Trifluormethyl)benzolsulfonylchlorid
Lieferant: Thermo Scientific
Synonyme:
3-(Trifluormethyl)benzol-1-sulfonylchlorid, 3-(Trifluormethyl)benzolsulfonsäurechlorid, 3-(Trifluormethyl)benzolsulfochlorid
BTB06459.1
BTB06459EA
MAYBBTB06459.1EA
0
EUR
MAYBBTB06459.1
MAYBBTB06459EA
3-(Trifluormethyl)benzolsulfonylchlorid
3-(Trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₄ClF₃O₂S Molecular Weight: 244,62 g/mol Siedepunkt: 75…76 °C (0,5 mmHg) Dichte: 1,52 g/cm³ (20 °C) |
MDL:
MFCD00014724 CAS-Nummer: 777-44-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...