1-tert-Butyl-3-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
3-tert-Butyliodobenzene, 3-tert-Butyliodbenzol, 1-(tert-Butyl)-3-iodobenzene
023858-1G
023858-5G
FLUO023858-1GEA
249
EUR
FLUO023858-1G
FLUO023858-5G
1-tert-Butyl-3-iodbenzol
1-tert-Butyl-3-iodbenzol
Formel:
C₁₀H₁₃I Molecular Weight: 260.12 g/mol |
MDL:
MFCD01764504 CAS-Nummer: 58164-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...