1-tert-Butyl-4-iodbenzol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-tert-Butyliodbenzol, 1-(tert-Butyl)-4-iodobenzene, 4-tert-Butyliodobenzene
L11603.03
L11603.06
L11603.03EA
49
EUR
L11603.03
L11603.06
1-tert-Butyl-4-iodbenzol ≥97%
1-tert-Butyl-4-iodbenzol
Purity: 97%
CAS No.: 35779-04-5
Molecular Formula: C10H13I
Formula Weight: 260.12
Flash Point: 106°(223°F)
Refractive Index: 1.5710
Boiling Point: 243°
Beilstein Reference: 1931810
MDL No.: MFCD00052339
Formel:
C₁₀H₁₃I Siedepunkt: 243 °C Dichte: 1,468 Flammpunkt: 106 °C (223 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052339 CAS-Nummer: 35779-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...