1-tert-Butyl-4-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
4-tert-Butyliodbenzol, 1-(tert-Butyl)-4-iodobenzene, 4-tert-Butyliodobenzene
CFF870.
008026-25G
008026-100G
CHMPCFF870.EA
0
EUR
CHMPCFF870.
FLUO008026-25G
FLUO008026-100G
1-tert-Butyl-4-iodbenzol
1-tert-Butyl-4-iodbenzol
Formel:
C₁₀H₁₃I Siedepunkt: 114 °C (0,05 mmHg) Dichte: 1,4392 g/cm³ (14 °C) |
MDL:
MFCD00052339 CAS-Nummer: 35779-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...