1-Ethyl-2-iodbenzol

Lieferant: CHEMPUR
Achtung

Synonyme: (o-Iodphenyl)ethan, 1-Iod-2-ethylbenzol, (2-Iodphenyl)ethan, 2-Ethyliodbenzol

FL1197.POR 001197-100G 001197-25G
CHMPFL1197.POREA 0 EUR
CHMPFL1197.POR FLUO001197-100G FLUO001197-25G
1-Ethyl-2-iodbenzol
1-Ethyl-2-iodbenzol
Formel: C₈H₉I
Molecular Weight: 232,06 g/mol
MDL: MFCD00040871
CAS-Nummer: 18282-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR