1-Ethyl-4-iodbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Ethyliodbenzol, (p-Iodphenyl)ethan

A14138.09 A14138.18
A14138.09EA 36.5 EUR
A14138.09 A14138.18
1-Ethyl-4-iodbenzol ≥98%
1-Ethyl-4-iodbenzol

Purity: 98% CAS No.: 25309-64-2 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 107°(224°F) Refractive Index: 1.5910 Odour: Mild Melting Point: -17° Boiling Point: 112-113°/20mm Beilstein Reference: 2076973 MDL No.: MFCD00060663

Formel: C₈H₉I
Molecular Weight: 232,06 g/mol
Siedepunkt: 112…113 °C (20 mmHg)
Schmelzpunkt: –17 °C
Dichte: 1,600
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00060663
CAS-Nummer: 25309-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR