(2-Iodphenyl)acetonitril 96%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Iodbenzylcyanid

L12767.03 L12767.06
L12767.03EA 27.3 EUR
L12767.03 L12767.06
(2-Iodphenyl)acetonitril 96%
(2-Iodphenyl)acetonitril

Purity: 96% CAS No.: 40400-15-5 Molecular Formula: C8H6IN Formula Weight: 243.05 Flash Point: 110°(230°F) Refractive Index: 1.6170 Boiling Point: 139-140°/13mm Beilstein Reference: 2083106 UN No.: 3276 MDL No.: MFCD00040888

Formel: C₈H₆IN
Molecular Weight: 243,05 g/mol
Siedepunkt: 139…140 °C (13 mmHg)
Dichte: 1,750
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040888
CAS-Nummer: 40400-15-5
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR