2'-Iodacetophenon ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(2-Iodphenyl)ethanon, o-Iodacetophenon, 1-(2-Iodphenyl)ethan-1-on

B25345.03 B25345.06
B25345.03EA 32.5 EUR
B25345.03 B25345.06
2'-Iodacetophenon ≥98%
2'-Iodacetophenon

Purity: 98+% CAS No.: 2142-70-3 Molecular Formula: C8H7IO Formula Weight: 246.05 Flash Point: 103°(218°F) Refractive Index: 1.6180 Boiling Point: 139-140°/12mm Beilstein Reference: 2325078 MDL No.: MFCD00094998

Formel: C₈H₇IO
Molecular Weight: 246,05 g/mol
Siedepunkt: 139…140 °C (12 mmHg)
Dichte: 1,746
Flammpunkt: 103 °C (218 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00094998
CAS-Nummer: 2142-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR