2'-Iodacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Iodphenyl)ethanon, o-Iodacetophenon, 1-(2-Iodphenyl)ethan-1-on

M-Clarity™ Qualitätsstufe = MQ200

540676-5G 540676-1G
SIAL540676-5GEA 0 EUR
SIAL540676-5G SIAL540676-1G
2'-Iodacetophenon, Sigma-Aldrich®
2'-Iodacetophenon
Formel: C₈H₇IO
Molecular Weight: 246,05 g/mol
MDL: MFCD00094998
CAS-Nummer: 2142-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR