4'-Iodacetophenon ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: p-Iodacetophenon, 1-(4-Iodphenyl)ethan-1-on, 1-(4-Iodphenyl)ethanon

A16794.06 A16794.14 A16794.22
A16794.06EA 34.4 EUR
A16794.06 A16794.14 A16794.22
4'-Iodacetophenon ≥98%
4'-Iodacetophenon

Purity: 98% CAS No.: 13329-40-3 Molecular Formula: C8H7IO Formula Weight: 246.05 Melting Point: 82-84° Beilstein Reference: 1857412

Formel: C₈H₇IO
Molecular Weight: 246,05 g/mol
Siedepunkt: 145…150 °C (20 mmHg)
Schmelzpunkt: 82…84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00045320
CAS-Nummer: 13329-40-3
EINECS: 236-372-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR