2-Iod-m-toluylsäure ≥98%

Lieferant: Thermo Scientific
Achtung

L19876.03 L19876.06
L19876.03EA 69.8 EUR
L19876.03 L19876.06
2-Iod-m-toluylsäure ≥98%
2-Iod-m-toluylsäure

Purity: 98% CAS No.: 108078-14-4 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 150-153° Beilstein Reference: 2439521

Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079764
CAS-Nummer: 108078-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR