4-Iodbenzylalkohol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Iodophenyl)methanol
L19278.03
L19278.06
L19278.03EA
40.2
EUR
L19278.03
L19278.06
4-Iodbenzylalkohol ≥97%
4-Iodbenzylalkohol
Purity: 97%
CAS No.: 18282-51-4
Molecular Formula: C7H7IO
Formula Weight: 234.04
Melting Point: 72-74°
Beilstein Reference: 1931621
Formel:
C₇H₇IO Molecular Weight: 234,04 g/mol Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01732720 CAS-Nummer: 18282-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...